3-Bromo-4-formylbenzonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-cyanobenzaldehyde

A188923-250MG A188923-1G
103816-144EA 41 USD
103816-144 103846-738 76974-786 77084-486
3-Bromo-4-formylbenzonitrile 97%
3-Bromo-4-formylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₄BrNO
MW: 210.027 g/mol
Melting Pt: 121…124 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09261031
CAS Number: 89891-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR