3-Bromo-4-fluoropyridine 98%

Supplier: Ambeed
Warning

A224402-100G A224402-10G A224402-5G
103819-108EA 21 USD
103819-108 103849-998 77000-014 77000-016 77088-652
3-Bromo-4-fluoropyridine 98%
3-Bromo-4-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrFN
MW: 175.9800 g/mol
Boiling Pt: 168.8 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114101
CAS Number: 116922-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR