3-Bromo-4-fluoropyridin-2-amine 95%

Supplier: Ambeed
Warning

A156478-100MG A156478-250MG
103804-890EA 736 USD
103804-890 103843-092 76984-866 76984-868
3-Bromo-4-fluoropyridin-2-amine 95%
3-Bromo-4-fluoropyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄BrFN₂
Boiling Pt: 234.8 °C (760 mm Hg)
Flash Pt: 95.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD26743631
CAS Number: 1417407-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR