3-Bromo-4-fluorophenol 97%
Supplier: Ambeed
|
Warning
|
A148536-100G
A148536-1G
A148536-25G
A148536-5G
103803-938EA
5
USD
103803-938
103842-054
77286-042
77317-514
77317-516
77317-518
3-Bromo-4-fluorophenol 97%
3-Bromo-4-fluorophenol
Precaución: for Research Use Only
Formula:
C₆H₄BrFO Boiling Pt: 240.5 °C (760 mm Hg) Flash Pt: 99.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03425884 CAS Number: 27407-11-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...