1-Bromo-2-fluoro-5-nitrobenzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromo-4-fluoronitrobenzene, 2-Bromo-1-fluoro-4-nitrobenzene
A137089-100G
A137089-10G
A137089-1000G
A137089-25G
A137089-5G
103808-554EA
353
USD
103808-554
77285-570
77285-572
77315-342
77315-344
77315-346
1-Bromo-2-fluoro-5-nitrobenzene 98%
1-Bromo-2-fluoro-5-nitrobenzene
Precaución: for Research Use Only
Formula:
C₆H₃BrFNO₂ MW: 220 g/mol Boiling Pt: 250.6 °C (760 mm Hg) Melting Pt: 55 °C Flash Pt: 105.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055433 CAS Number: 701-45-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...