3-Bromo-4-fluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A110765-100G A110765-10G A110765-1G A110765-25G
103823-532EA 6 USD
103823-532 76765-788 76765-790 76765-792 76765-794
3-Bromo-4-fluorobenzaldehyde 98%
3-Bromo-4-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrFO
MW: 203.01 g/mol
Boiling Pt: 138…139 °C (760 mm Hg)
Melting Pt: 31…33 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00042186
CAS Number: 77771-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR