2-Bromo-1-fluoro-4-methoxybenzene 95%

Supplier: Ambeed
Warning

A152112-100G A152112-10G A152112-1G A152112-5G A152112-25G
103804-350EA 5 USD
103804-350 103842-506 76812-258 76812-260 77299-924 77299-926 77318-116
2-Bromo-1-fluoro-4-methoxybenzene 95%
2-Bromo-1-fluoro-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD13185359
CAS Number: 1161497-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR