3-Bromo-4-fluoroaniline 98%
Supplier: Ambeed
|
Warning
|
A159788-100G
A159788-10G
A159788-25G
A159788-5G
A159788-500G
103843-514EA
5
USD
103843-514
76812-658
76812-660
77319-418
77319-420
77883-253
3-Bromo-4-fluoroaniline 98%
3-Bromo-4-fluoroaniline
Precaución: for Research Use Only
Formula:
C₆H₅BrFN Boiling Pt: 235 °C (760 mm Hg) Melting Pt: 34…36 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788559 CAS Number: 656-64-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...