3'-Bromo-4'-fluoroacetophenone 98%

Supplier: Ambeed
Warning

A147307-100G A147307-10G A147307-1G A147307-500G A147307-25G A147307-5G
103841-858EA 6 USD
103841-858 77235-254 77235-256 77235-258 77235-260 77317-226 77317-228
3'-Bromo-4'-fluoroacetophenone 98%
3'-Bromo-4'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆BrFO
MW: 217.04 g/mol
Melting Pt: 52…57 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00042466
CAS Number: 1007-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR