3-Bromo-4-ethyl-5-fluoropyridine 95%

Supplier: Ambeed
Warning

A135339-1G A135339-100MG
103802-134EA 304 USD
103802-134 77298-768 77885-156
3-Bromo-4-ethyl-5-fluoropyridine 95%
3-Bromo-4-ethyl-5-fluoropyridine

Precaución: for Research Use Only

Formula: C7H7BrFN
Storage Temperature: Refrigerator
MDL Number: MFCD23140982
CAS Number: 1374655-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR