3-Bromoisonicotinonitrile 97%

Supplier: Ambeed
Warning

A353184-1G A353184-25G A353184-5G A353184-100G A353184-100MG A353184-250MG
103826-086EA 14 USD
103826-086 103837-088 76840-202 76840-204 76840-206 77003-284 77003-286 77003-288
3-Bromoisonicotinonitrile 97%
3-Bromoisonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₃BrN₂
MW: 183.01 g/mol
Boiling Pt: 267.5 °C (760 mm Hg)
Flash Pt: 115.568 °C
Storage Temperature: Ambient
MDL Number: MFCD00234143
CAS Number: 13958-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR