3-Bromo-4-chloroquinoline 97%

Supplier: Ambeed
Danger

A432083-1G A432083-5G A432083-250MG
103827-530EA 46 USD
103827-530 103838-596 76842-288 76842-290 77116-066
3-Bromo-4-chloroquinoline 97%
3-Bromo-4-chloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
Melting Pt: 66…67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05738886
CAS Number: 74575-17-0
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR