3-Bromo-4-chloropyridin-2-amine 98%

Supplier: Ambeed
Warning

A160126-100MG A160126-10G A160126-250MG A160126-1G A160126-5G
103805-336EA 81 USD
103805-336 103843-566 76812-676 76812-678 76812-680 77300-438 77300-440
3-Bromo-4-chloropyridin-2-amine 98%
3-Bromo-4-chloropyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄BrClN₂
MW: 207.46 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15143381
CAS Number: 221297-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR