3-Bromo-4-chlorophenol 98%

Supplier: Ambeed
Warning

A121796-100G A121796-10G A121796-25G A121796-1G A121796-5G
103800-134EA 9 USD
103800-134 77271-552 77271-554 77271-556 77312-264 77312-266
3-Bromo-4-chlorophenol 98%
3-Bromo-4-chlorophenol

Precaución: for Research Use Only

Formula: C₆H₄BrClO
Boiling Pt: 273 °C (760 mm Hg)
Flash Pt: 119 °C
Storage Temperature: Ambient
MDL Number: MFCD00070740
CAS Number: 13659-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR