1-Bromo-2-chloro-5-nitrobenzene 98%

Supplier: Ambeed
Warning

A107699-10G A107699-25G
103823-000EA 6 USD
103823-000 77148-544 77148-546
1-Bromo-2-chloro-5-nitrobenzene 98%
1-Bromo-2-chloro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
Boiling Pt: 287.2 °C (760 mm Hg)
Melting Pt: 58 °C
Flash Pt: 127.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00100437
CAS Number: 16588-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR