3-Bromo-4-chlorobenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A128341-100G
A128341-10G
A128341-1G
A128341-5G
A128341-25G
103801-110EA
6
USD
103801-110
76782-148
76782-150
77298-292
77298-294
77313-420
3-Bromo-4-chlorobenzaldehyde 98%
3-Bromo-4-chlorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₄BrClO MW: 219.4600 g/mol Melting Pt: 70 °C Flash Pt: 121.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08059100 CAS Number: 86265-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...