3-Bromo-4-chloroaniline 98%

Supplier: Ambeed
Warning

A278990-25G A278990-5G A278990-100G A278990-10G A278990-500G A278990-1G
103813-446EA 5 USD
103813-446 103853-390 77093-316 77093-318 77276-924 77276-926 77276-928 77305-314
3-Bromo-4-chloroaniline 98%
3-Bromo-4-chloroaniline

Precaución: for Research Use Only

Formula: C₆H₅BrClN
Boiling Pt: 290.6 °C (760 mm Hg)
Melting Pt: 82 °C
Flash Pt: 129.535 °C
Storage Temperature: Ambient
MDL Number: MFCD00173905
CAS Number: 823-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR