3-Bromo-4-chloro-5-methylpyridine 95%

Supplier: Ambeed
Warning

A129085-100MG A129085-250MG A129085-1G
103801-226EA 117 USD
103801-226 77234-404 77234-406 77313-592
3-Bromo-4-chloro-5-methylpyridine 95%
3-Bromo-4-chloro-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClN
Boiling Pt: 233.5±35.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD18415475
CAS Number: 1261786-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR