3-Bromo-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one, 3-Bromo-2,3,4,5-tetrahydro-2H-benzo[b]azepin-2-one, 3-Bromo-1,3,4,5-tetrahydro-2H-1-benzazepin-2-one

A295481-100G A295481-10G A295481-500G A295481-25G A295481-5G
103835-194EA 7 USD
103835-194 76648-042 76648-044 76648-046 76837-852 76837-854
3-Bromo-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one 97%
3-Bromo-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one

Precaución: for Research Use Only

Formula: C10H10BrNO
MW: 240.1000 g/mol
Boiling Pt: 383.2 °C (760 mm Hg)
Melting Pt: 164 °C
Flash Pt: 185.542 °C
Storage Temperature: Ambient
MDL Number: MFCD03085918
CAS Number: 86499-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR