3-Bromo-4-(trifluoromethyl)benzoic acid 95%

Supplier: Ambeed
Warning

A330850-25G A330850-250MG
103825-566EA 16 USD
103825-566 103836-528 76839-512 76978-236
3-Bromo-4-(trifluoromethyl)benzoic acid 95%
3-Bromo-4-(trifluoromethyl)benzoic acid

Precaución: For research use only

Formula: C₈H₄BrF₃O₂
Boiling Pt: 292.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD13185386
CAS Number: 581813-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR