3-Bromo-4-(trifluoromethyl)benzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-formylbenzotrifluoride, 3-Bromo-α,α,α-trifluoro-p-tolualdehyde

A247420-100MG A247420-10G A247420-250MG A247420-1G A247420-5G
103811-768EA 63 USD
103811-768 103851-602 76874-544 76874-546 76874-548 77090-940 77090-942
3-Bromo-4-(trifluoromethyl)benzaldehyde 98%
3-Bromo-4-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O
MW: 253.016 g/mol
Boiling Pt: 244.6±40.0 °C (760 mm Hg)
Flash Pt: 101.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13185384
CAS Number: 372120-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR