3-Bromo-4-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

A340899-1G A340899-25G A340899-100G A340899-10G A340899-250MG A340899-5G
103825-816EA 17 USD
103825-816 103836-794 76839-810 76839-812 76978-400 76978-402 76978-404 77306-526
3-Bromo-4-(trifluoromethyl)aniline 98%
3-Bromo-4-(trifluoromethyl)aniline

Precaución: For research use only

Formula: C₇H₅BrF₃N
MW: 240.0200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12828280
CAS Number: 172215-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR