3-Bromo-2-nitrobenzaldehyde 97%

Supplier: Ambeed
Warning

A214850-10G A214850-250MG
103818-436EA 176 USD
103818-436 103849-138 77890-275 77890-276
3-Bromo-2-nitrobenzaldehyde 97%
3-Bromo-2-nitrobenzaldehyde

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₇H₄BrNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00462872
CAS Number: 882772-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR