3-Bromo-2-methylpyridine 98%
Supplier: Ambeed
|
Warning
|
A161031-100G
A161031-10G
A161031-500G
A161031-1G
A161031-25G
A161031-5G
103805-442EA
5
USD
103805-442
103843-680
77273-290
77273-292
77273-294
77319-640
77319-642
77319-644
3-Bromo-2-methylpyridine 98%
3-Bromo-2-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₆BrN Boiling Pt: 179.6 °C (760 mm Hg) Flash Pt: 62.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191224 CAS Number: 38749-79-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...