3-Bromo-o-cresol 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-methylphenol

A180823-10G A180823-250MG A180823-1G A180823-5G
103807-564EA 20 USD
103807-564 103845-926 76974-454 76974-456 77083-436 77083-438
3-Bromo-o-cresol 98%
3-Bromo-o-cresol

Precaución: for Research Use Only

Formula: C₇H₇BrO
MW: 187.0339 g/mol
Melting Pt: 92…96 °C
Storage Temperature: Ambient
MDL Number: MFCD11100990
CAS Number: 7766-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR