3-Bromo-2-methylimidazo[1,2-b]pyridazine 97%

Supplier: Ambeed
Warning

A201104-100MG A201104-250MG
103817-266EA 187 USD
103817-266 77237-162 77237-164
3-Bromo-2-methylimidazo[1,2-b]pyridazine 97%
3-Bromo-2-methylimidazo[1,2-b]pyridazine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22038943
CAS Number: 1368313-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR