3-Bromo-2-methylbenzo[b]thiophene 97%

Supplier: Ambeed
Warning

A132431-100MG A132431-250MG A132431-1G
103801-736EA 43 USD
103801-736 77272-064 77272-066 77298-558
3-Bromo-2-methylbenzo[b]thiophene 97%
3-Bromo-2-methylbenzo[b]thiophene

Precaución: for Research Use Only

Formula: C9H7BrS
Storage Temperature: Refrigerator
MDL Number: MFCD01830303
CAS Number: 10243-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR