3-Bromo-2-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

A252133-100MG A252133-10G A252133-250MG A252133-1G A252133-25G A252133-5G
103812-028EA 17 USD
103812-028 103851-870 76815-678 76815-680 76815-682 77091-274 77091-276 77091-278
3-Bromo-2-methylbenzaldehyde 97%
3-Bromo-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO
Boiling Pt: 255.3 °C (760 mm Hg)
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11847533
CAS Number: 83647-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR