3-Bromo-o-toluidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-6-bromotoluene, 3-Bromo-2-methylaniline

A106347-10G A106347-25G A106347-1G A106347-5G A106347-100G A106347-500G
103797-026EA 9 USD
103797-026 103822-778 77025-462 77025-464 77283-466 77309-636 77549-042 77570-374
3-Bromo-o-toluidine 98%
3-Bromo-o-toluidine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 257 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00051579
CAS Number: 55289-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR