3-Bromo-2-methyl-5-nitropyridine 98%

Supplier: Ambeed
Warning

A112374-10G A112374-25G A112374-1G A112374-5G
103798-004EA 10 USD
103798-004 103823-838 77094-046 77094-048 77284-228 77310-646
3-Bromo-2-methyl-5-nitropyridine 98%
3-Bromo-2-methyl-5-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂
Boiling Pt: 270.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08688606
CAS Number: 186593-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR