3-Bromo-2-methoxypyridine 98%

Supplier: Ambeed
Warning

A197382-1G A197382-100G A197382-10G A197382-25G A197382-500G
103816-914EA 8 USD
103816-914 103847-536 77085-540 77112-028 77112-030 77112-032 77685-493
3-Bromo-2-methoxypyridine 98%
3-Bromo-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.02 g/mol
Boiling Pt: 190.387 °C (760 mm Hg)
Flash Pt: 68.944 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095349
CAS Number: 13472-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR