3-Bromo-2-methoxybenzoic acid 98%

Supplier: Ambeed
Danger

A174160-1G A174160-25G A174160-5G A174160-10G
103806-924EA 10 USD
103806-924 103845-222 77082-510 77082-512 77082-514 77273-814
3-Bromo-2-methoxybenzoic acid 98%
3-Bromo-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
MW: 231.05 g/mol
Boiling Pt: 327.6 °C (760 mm Hg)
Melting Pt: 121…123 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00045840
CAS Number: 101084-39-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR