3-Bromo-2-methoxyaniline 98%

Supplier: Ambeed
Danger

Synonyms: 3-Bromo-o-anisidine, 2-Amino-6-bromoanisole

A123251-100G A123251-10G A123251-250MG A123251-25G A123251-1G A123251-5G
103800-372EA 13 USD
103800-372 76870-348 76870-350 76870-352 76870-354 77312-488 77312-490
3-Bromo-2-methoxyaniline 98%
3-Bromo-2-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.05 g/mol
Boiling Pt: 267.9 °C (760 mm Hg)
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD13186737
CAS Number: 116557-46-1
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR