3-Bromo-2-methoxy-5-methylpyridine 98%

Supplier: Ambeed
Warning

A112777-250MG A112777-1G A112777-5G
103798-104EA 8 USD
103798-104 103823-940 76781-220 77310-762 77310-764
3-Bromo-2-methoxy-5-methylpyridine 98%
3-Bromo-2-methoxy-5-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.049 g/mol
Boiling Pt: 219.6±35.0 °C (760 mm Hg)
Flash Pt: 86.6 °C
Storage Temperature: Ambient
MDL Number: MFCD08061306
CAS Number: 717843-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR