3-Bromo-2-methoxy-4-picoline 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-methoxy-4-methylpyridine

A108458-10G A108458-250MG A108458-5G A108458-1G
103797-350EA 22 USD
103797-350 103823-130 76869-578 76869-580 77283-746 77309-970
3-Bromo-2-methoxy-4-picoline 97%
3-Bromo-2-methoxy-4-picoline

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.049 g/mol
Boiling Pt: 223.4±35.0 °C (760 mm Hg)
Flash Pt: 88.9 °C
Storage Temperature: Ambient
MDL Number: MFCD08061305
CAS Number: 717843-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR