3-Bromo-2-hydroxybenzoic acid 98%

Supplier: Ambeed
Warning

A362033-1G A362033-250MG
103826-296EA 7 USD
103826-296 103837-310 76840-524 77115-314
3-Bromo-2-hydroxybenzoic acid 98%
3-Bromo-2-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C7H5BrO3
Boiling Pt: 301.4 °C (760 mm Hg)
Flash Pt: 136.1 °C
Storage Temperature: Ambient
MDL Number: MFCD03265705
CAS Number: 3883-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR