3-Bromo-2-formylpyridine 97%
Supplier: Ambeed
|
Warning
|
A139347-10G
A139347-250MG
A139347-25G
A139347-1G
A139347-5G
103802-728EA
23
USD
103802-728
77272-414
77272-416
77272-418
77315-812
77315-814
3-Bromo-2-formylpyridine 97%
3-Bromo-2-formylpyridine
Precaución: for Research Use Only
Formula:
C₆H₄BrNO Storage Temperature: Refrigerator |
MDL Number:
MFCD07781220 CAS Number: 405174-97-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...