3-Bromo-2-fluoropyridine 98%

Supplier: Ambeed
Warning

A138617-100G A138617-10G A138617-500G A138617-1G A138617-25G A138617-5G
103802-638EA 9 USD
103802-638 76972-476 76972-478 76972-480 77299-002 77315-678 77315-680
3-Bromo-2-fluoropyridine 98%
3-Bromo-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrFN
Boiling Pt: 179.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04112496
CAS Number: 36178-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR