3-Bromo-2-fluorophenol 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2-fluoro-3-hydroxybenzene

A191308-10G A191308-250MG A191308-1G A191308-5G
103816-364EA 27 USD
103816-364 103846-966 76784-930 76784-932 77084-798 77084-800
3-Bromo-2-fluorophenol 98%
3-Bromo-2-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₄BrFO
Boiling Pt: 207.4 °C (760 mm Hg)
Melting Pt: 52…54 °C
Flash Pt: 79.2 °C
Storage Temperature: Ambient
MDL Number: MFCD09864673
CAS Number: 156682-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR