3-Bromo-α,α,α,2-tetrafluorotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-fluorobenzotrifluoride, 1-Bromo-2-fluoro-3-(trifluoromethyl)benzene

A619680-5G A619680-100G A619680-10G A619680-1G A619680-25G
103840-206EA 24 USD
103840-206 76844-380 77117-450 77117-452 77307-960 77307-962
3-Bromo-α,α,α,2-tetrafluorotoluene 98%
3-Bromo-α,α,α,2-tetrafluorotoluene

Precaución: for Research Use Only

Formula: C₇H₃BrF₄
MW: 243 g/mol
Boiling Pt: 164.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00070812
CAS Number: 144584-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR