3-Bromo-2-fluorobenzoic acid 98%

Supplier: Ambeed
Warning

A243096-25G A243096-5G A243096-100G A243096-10G A243096-500G
103811-502EA 5 USD
103811-502 103851-326 77090-516 77090-518 77276-084 77276-086 77276-088
3-Bromo-2-fluorobenzoic acid 98%
3-Bromo-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrFO₂
Boiling Pt: 298.2 °C (760 mm Hg)
Melting Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00665763
CAS Number: 161957-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR