3-Bromo-2-fluorobenzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-fluorobenzenecarbaldehyde

A326346-25G A326346-5G A326346-100G A326346-10G A326346-500G A326346-1G
103825-428EA 6 USD
103825-428 103836-376 76839-330 76839-332 76978-168 76978-170 76978-172 77306-336
3-Bromo-2-fluorobenzaldehyde 98%
3-Bromo-2-fluorobenzaldehyde

Precaución: For research use only

Formula: C₇H₄BrFO
Boiling Pt: 233.6 °C (760 mm Hg)
Melting Pt: 43…46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09027089
CAS Number: 149947-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR