3-Bromo-2-fluoro-5-methylpyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromo-2-fluoro-5-picoline
A138994-100G
A138994-10G
A138994-250MG
A138994-1G
A138994-25G
A138994-5G
103802-684EA
6
USD
103802-684
76996-796
76996-798
76996-800
77315-750
77315-752
77315-754
3-Bromo-2-fluoro-5-methylpyridine 98%
3-Bromo-2-fluoro-5-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₅BrFN MW: 190.01 Boiling Pt: 207.8 °C (760 mm Hg) Flash Pt: 79.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095305 CAS Number: 17282-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...