3-Bromo-2-chlorothiophene 97%

Supplier: Ambeed
Warning

A146101-25G A146101-5G A146101-10G
103803-628EA 5 USD
103803-628 103841-714 77317-020 77317-022 77748-022
3-Bromo-2-chlorothiophene 97%
3-Bromo-2-chlorothiophene

Precaución: for Research Use Only

Formula: C₄H₂BrClS
MW: 197.48 g/mol
Boiling Pt: 197.3 °C (760 mm Hg)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00043883
CAS Number: 40032-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR