4-Amino-3-bromo-2-chloropyridine 97%

Supplier: Ambeed
Danger

A257682-10G A257682-250MG A257682-1G A257682-25G A257682-5G
103812-330EA 14 USD
103812-330 103852-198 77000-872 77000-874 77091-716 77091-718 77091-720
4-Amino-3-bromo-2-chloropyridine 97%
4-Amino-3-bromo-2-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₄BrClN₂
Melting Pt: 148…153 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09263478
CAS Number: 215364-85-5
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR