3-Bromo-2-chlorophenol 98%

Supplier: Ambeed
Warning

A213231-1G A213231-25G A213231-5G A213231-100G A213231-100MG A213231-10G A213231-250MG
103818-270EA 8 USD
103818-270 103848-976 77087-464 77087-466 77087-468 77112-508 77112-510 77112-512 77112-514
3-Bromo-2-chlorophenol 98%
3-Bromo-2-chlorophenol

Precaución: for Research Use Only

Formula: C₆H₄BrClO
MW: 207.452 g/mol
Boiling Pt: 246.1 °C (760 mm Hg)
Flash Pt: 102.7 °C
Storage Temperature: Ambient
MDL Number: MFCD08166321
CAS Number: 863870-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR