3-Bromo-2-chlorobenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A194387-10G
A194387-250MG
A194387-1G
A194387-25G
A194387-5G
103816-648EA
11
USD
103816-648
103847-258
76974-998
76975-000
77302-416
77302-418
77302-420
3-Bromo-2-chlorobenzoic acid 95%
3-Bromo-2-chlorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₄BrClO₂ Melting Pt: 168…169 °C Flash Pt: 157.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD01569399 CAS Number: 56961-27-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...