3-Bromo-2-chloro-5-nitropyridine 98%

Supplier: Ambeed
Warning

A242324-100G A242324-10G A242324-25G A242324-5G A242324-1G
103811-464EA 10 USD
103811-464 103851-276 76976-508 76976-510 77090-448 77090-450 77304-244
3-Bromo-2-chloro-5-nitropyridine 98%
3-Bromo-2-chloro-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClN₂O₂
Boiling Pt: 293.8 °C (760 mm Hg)
Melting Pt: 54…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00233989
CAS Number: 5470-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR