3-Bromo-2-chloro-5-iodopyridine 97%

Supplier: Ambeed
Warning

A102906-100MG A102906-250MG A102906-1G
103796-444EA 29 USD
103796-444 103822-146 76821-310 76821-312 77309-016
3-Bromo-2-chloro-5-iodopyridine 97%
3-Bromo-2-chloro-5-iodopyridine

Precaución: for Research Use Only

Formula: C5H2BrClIN
Boiling Pt: 304.7±42.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD13185354
CAS Number: 1211586-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR