3-Bromo-2-chloro-5-fluoropyridine 95%
Supplier: Ambeed
|
Warning
|
A123345-10G
A123345-250MG
A123345-1G
A123345-5G
A123345-25G
103800-394EA
10
USD
103800-394
76870-356
76870-358
77312-510
77312-512
77592-406
3-Bromo-2-chloro-5-fluoropyridine 95%
3-Bromo-2-chloro-5-fluoropyridine
Precaución: for Research Use Only
Formula:
C₅H₂BrClFN Boiling Pt: 202.6 °C (760 mm Hg) Flash Pt: 76.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03095269 CAS Number: 884494-36-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...